ENA311759136 Enamine

1H,2H,3H,4H,5H-thieno[3,2-e][1,4]diazepin-5-one

  • MDL number MFCD17977149
  • Cas Number : 304021-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71489
Availability details for
Quantity
  • Availability
  • Add to Cart