ENA311759144 Enamine

5-(5-bromo-2-chlorophenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD02664099
  • Cas Number : 299442-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69517
Availability details for
Quantity
  • Availability
  • Add to Cart