ENA311759148 Enamine

4-phenylbutanethioamide

  • MDL number MFCD09809278
  • Cas Number : 177665-17-7
  • Molecular Weight : 179.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69585
Availability details for
Quantity
  • Availability
  • Add to Cart