ENA311759155 Enamine

5-methylidene-2,3,4,5-tetrahydro-1-benzoxepine

  • MDL number MFCD08706332
  • Cas Number : 20426-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69728

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69728
Availability details for
Quantity
  • Availability
  • Add to Cart