ENA311759156 Enamine

methyl 3-amino-3-(2-methylphenyl)propanoate hydrochloride

  • MDL number MFCD06740364
  • Cas Number : 1258651-74-9
  • Molecular Weight : 229.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69731
Availability details for
Quantity
  • Availability
  • Add to Cart