ENA311759160 Enamine

1-(2-bromoethoxy)-4-(trifluoromethyl)benzene

  • MDL number MFCD10690616
  • Cas Number : 125174-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69793

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69793
Availability details for
Quantity
  • Availability
  • Add to Cart