ENA311759162 Enamine

4-methyl-1,3-thiazole-5-carbohydrazide

  • MDL number MFCD09815062
  • Cas Number : 89179-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-167
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69800

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69800
Availability details for
Quantity
  • Availability
  • Add to Cart