ENA311759167 Enamine

6-bromo-3-methyl-3,4-dihydro-2H-1-benzothiopyran-4-one

  • MDL number MFCD12132772
  • Cas Number : 1153834-53-7
  • Molecular Weight : 257.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69817
Availability details for
Quantity
  • Availability
  • Add to Cart