ENA311759170 Enamine

2-chloro-N-[1-(naphthalen-1-yl)ethyl]propanamide

  • MDL number MFCD11546999
  • Cas Number : 1094533-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28053
Availability details for
Quantity
  • Availability
  • Add to Cart