ENA311759176 Enamine

1H,2H,4H-thieno[3,2-d][1,3]oxazine-2,4-dione

  • MDL number MFCD00106648
  • Cas Number : 78756-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69862

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-69862
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart