ENA311759193 Enamine

2-iodopyrimidine

  • MDL number MFCD09835340
  • Cas Number : 31462-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 28-32
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69951
Availability details for
Quantity
  • Availability
  • Add to Cart