ENA311759196 Enamine

3-cyclopentyl-3-oxo-2-phenylpropanenitrile

  • MDL number MFCD11208540
  • Cas Number : 1094230-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69955
Availability details for
Quantity
  • Availability
  • Add to Cart