ENA311759205 Enamine

2-[2-(4-chlorophenyl)-1,3-oxazol-4-yl]acetic acid

  • MDL number MFCD11587874
  • Cas Number : 22086-88-0
  • Molecular Weight : 237.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69974
Availability details for
Quantity
  • Availability
  • Add to Cart