ENA311759207 Enamine

2-chloro-N-[(3-propoxyphenyl)methyl]acetamide

  • MDL number MFCD16495550
  • Cas Number : 1226036-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-48490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-48490
Availability details for
Quantity
  • Availability
  • Add to Cart