ENA311759212 Enamine

2-oxo-6-(2-phenylethyl)-1,2,5,6,7,8-hexahydro-1,6-naphthyridine-3-carbonitrile

  • MDL number MFCD17977006
  • Cas Number : 1258651-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69985

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69985
Availability details for
Quantity
  • Availability
  • Add to Cart