ENA311759214 Enamine

1-(thiophen-3-yl)propan-2-one

  • MDL number MFCD09834575
  • Cas Number : 62119-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69987
Availability details for
Quantity
  • Availability
  • Add to Cart