ENA311759217 Enamine

2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-2-methylpropanenitrile

  • MDL number MFCD12792057
  • Cas Number : 1240384-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69991
Availability details for
Quantity
  • Availability
  • Add to Cart