ENA311759219 Enamine

2-(1,4-diazepan-1-yl)ethan-1-ol

  • MDL number MFCD01565842
  • Cas Number : 53427-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69993
Availability details for
Quantity
  • Availability
  • Add to Cart