ENA311759221 Enamine

3-methyl-3-phenylcyclobutan-1-one

  • MDL number MFCD17977008
  • Cas Number : 151990-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69995

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69995
Availability details for
Quantity
  • Availability
  • Add to Cart