ENA311759228 Enamine

6-chloro-N-(1,3-thiazol-2-yl)pyridine-3-sulfonamide

  • MDL number MFCD04176837
  • Cas Number : 622804-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70006

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70006
Availability details for
Quantity
  • Availability
  • Add to Cart