ENA311759233 Enamine

[(2,5-dimethoxyphenyl)methyl](methyl)amine hydrochloride

  • MDL number MFCD15201659
  • Cas Number : 1158286-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70014
Availability details for
Quantity
  • Availability
  • Add to Cart