ENA311759240 Enamine

[2-(dimethylamino)ethyl](2-methoxyethyl)amine

  • MDL number MFCD11154689
  • Cas Number : 819813-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70021

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70021
Availability details for
Quantity
  • Availability
  • Add to Cart