ENA311759247 Enamine

1-(4-tert-butyl-1,3-thiazol-2-yl)methanamine

  • MDL number MFCD05222069
  • Cas Number : 184839-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70044
Availability details for
Quantity
  • Availability
  • Add to Cart