ENA311759248 Enamine

2-(4-tert-butyl-1,3-thiazol-2-yl)ethan-1-amine

  • MDL number MFCD05221351
  • Cas Number : 642081-26-3
  • Molecular Weight : 184.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70045
Availability details for
Quantity
  • Availability
  • Add to Cart