ENA311759251 Enamine

4-oxo-2,3,4,5-tetrahydro-1H-1,5-benzodiazepine-7-carboxamide

  • MDL number MFCD17977020
  • Cas Number : 224811-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70057

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70057
Availability details for
Quantity
  • Availability
  • Add to Cart