ENA311759253 Enamine

5-fluoro-2-(2-methyl-1H-imidazol-1-yl)aniline

  • MDL number MFCD04971021
  • Cas Number : 869942-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153...155
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70059

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70059
Availability details for
Quantity
  • Availability
  • Add to Cart