ENA311759267 Enamine

4-hydroxy-1-(2,4,6-trichlorophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11523851
  • Cas Number : 1152602-90-8
  • Molecular Weight : 307.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70084
Availability details for
Quantity
  • Availability
  • Add to Cart