ENA311759274 Enamine

1,1,1,3,4,4,5,5,5-nonafluoro-2-(trifluoromethyl)pent-2-ene

  • MDL number MFCD00015724
  • Cas Number : 1584-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point -12
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70091

Safety Information

signalword Danger
safety_statement 16-29-33
msds https://www.enaminestore.com/catalog/EN300-70091
pictograms GHS02
Availability details for
Quantity
  • Availability
  • Add to Cart