ENA311759278 Enamine

1-(6-methoxypyridin-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD11149187
  • Cas Number : 138949-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70096
Availability details for
Quantity
  • Availability
  • Add to Cart