ENA311759279 Enamine

3-benzyl-3-azabicyclo[3.1.1]heptan-6-one

  • MDL number MFCD21642063
  • Cas Number : 1240529-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-74
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70097
Availability details for
Quantity
  • Availability
  • Add to Cart