ENA311759281 Enamine

1-(3-bromopropyl)-3-nitro-1H-pyrazole

  • MDL number MFCD14594889
  • Cas Number : 1250489-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35...37
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70099

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70099
Availability details for
Quantity
  • Availability
  • Add to Cart