ENA311759282 Enamine

N-[(4-chlorophenyl)methyl]-1H-imidazole-1-carboxamide

  • MDL number MFCD11099712
  • Cas Number : 149047-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70100

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70100
Availability details for
Quantity
  • Availability
  • Add to Cart