ENA311759285 Enamine

N-(2-aminoethyl)-N-methylcyclopentanamine

  • MDL number MFCD09754454
  • Cas Number : 859340-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70103
Availability details for
Quantity
  • Availability
  • Add to Cart