ENA311759287 Enamine

4-bromo-2-(bromomethyl)-1-(pentyloxy)benzene

  • MDL number MFCD11180444
  • Cas Number : 1094273-06-9
  • Molecular Weight : 336.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70105
Availability details for
Quantity
  • Availability
  • Add to Cart