ENA311759288 Enamine

1-phenyloctane-1,3-dione

  • MDL number MFCD11178609
  • Cas Number : 55846-68-9
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70106
Availability details for
Quantity
  • Availability
  • Add to Cart