ENA311759291 Enamine

4-aminophenyl tert-butyl carbonate

  • MDL number MFCD18089381
  • Cas Number : 95932-39-1
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70109
Availability details for
Quantity
  • Availability
  • Add to Cart