ENA311759295 Enamine

furan-3-carboxamide

  • MDL number MFCD02179221
  • Cas Number : 609-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70113
Availability details for
Quantity
  • Availability
  • Add to Cart