ENA311759297 Enamine

1-butyl-4-isocyanatocyclohexane

  • MDL number MFCD09928695
  • Cas Number : 1016692-78-6
  • Molecular Weight : 181.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70116

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P272,P273,P280,P285,P301+P310,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P304+P341,P305+P351+P338,P310,P311,P312,P330,P333+P313,P342+P311,P361,P363,P391,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-70116
pictograms GHS05,GHS06,GHS08,GHS09
hstatements H301,H311,H314,H317,H331,H334,H335,H411
Availability details for
Quantity
  • Availability
  • Add to Cart