ENA311759301 Enamine

6-(2-bromopropanoyl)-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD11103549
  • Cas Number : 62618-72-8
  • Molecular Weight : 282.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70127
Availability details for
Quantity
  • Availability
  • Add to Cart