ENA311759305 Enamine

4-hydrazino-5,6-dimethyl-2-pyridin-4-ylpyrimidine

  • MDL number MFCD16727075
  • Cas Number : 1247850-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70131

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70131
Availability details for
Quantity
  • Availability
  • Add to Cart