ENA311759308 Enamine

1-amino-3-(3,4-difluorophenoxy)propan-2-ol

  • MDL number MFCD12137007
  • Cas Number : 1157091-16-1
  • Molecular Weight : 203.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70134
Availability details for
Quantity
  • Availability
  • Add to Cart