ENA311759329 Enamine

2-phenylpyrimidine-4-carbaldehyde

  • MDL number MFCD10696995
  • Cas Number : 1014-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70208
Availability details for
Quantity
  • Availability
  • Add to Cart