ENA311759331 Enamine

[5-(4-methoxyphenyl)-1,2,4-oxadiazol-3-yl]methanol

  • MDL number MFCD09027670
  • Cas Number : 1258639-51-8
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70210
Availability details for
Quantity
  • Availability
  • Add to Cart