ENA311759333 Enamine

4-cyclopropyl-5-(piperidin-4-yl)-4H-1,2,4-triazole-3-thiol hydrochloride

  • MDL number MFCD15200708
  • Cas Number : 1158449-20-7
  • Molecular Weight : 260.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70212
Availability details for
Quantity
  • Availability
  • Add to Cart