ENA311759338 Enamine

4-[(2-chlorophenyl)methyl]-1-methylpiperidin-4-ol

  • MDL number MFCD12766274
  • Cas Number : 1183059-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70218

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70218
Availability details for
Quantity
  • Availability
  • Add to Cart