ENA311759339 Enamine

4-(2-oxo-2,3-dihydro-1H-indol-5-yl)butanoic acid

  • MDL number MFCD11168389
  • Cas Number : 1042520-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70222

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70222
Availability details for
Quantity
  • Availability
  • Add to Cart