ENA311759340 Enamine

1-(5,6-dichloropyridine-3-carbonyl)pyrrolidine-2-carboxamide

  • MDL number MFCD11538680
  • Cas Number : 1132702-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70227
Availability details for
Quantity
  • Availability
  • Add to Cart