ENA311759345 Enamine

1-acetylpiperidine-3-sulfonyl chloride

  • MDL number MFCD14688480
  • Cas Number : 1251248-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55114

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55114
Availability details for
Quantity
  • Availability
  • Add to Cart