ENA311759347 Enamine

2'-methyl-[1,1'-biphenyl]-3-carbaldehyde

  • MDL number MFCD03424623
  • Cas Number : 371764-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70235
Availability details for
Quantity
  • Availability
  • Add to Cart