ENA311759349 Enamine

3-(4-aminophenoxy)-N-methylpropanamide

  • MDL number MFCD12790844
  • Cas Number : 1225702-73-7
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70237
Availability details for
Quantity
  • Availability
  • Add to Cart